Order Entry
Ireland
ContactUsLinkComponent
Biotin-PEG4-NHS biodetection
Irritant
Biotin-PEG4-NHS biodetection
Catalog # AATB3022
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Biotin-PEG4-NHS biodetection
Catalog # AATB3022
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :588.67 g/mol
ADR:5,II
 :GHS07
CAS Number:459426-22-3
 :MFCD08458915
Formula:C₂₅H₄₀N₄O₁₀S
MDL Number:MFCD08457831