
Biotin C2 maleimide biodetection
Catalog # AATB3005
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:366.3 g/mol
ADR:5,II
:GHS07
CAS Number:139554-72-6
Formula:C₁₆H₂₂SN₄O₄
MDL Number:MFCD02094223