Fritted Spin Column, 3 ml
Catalog # GENO786-1858
Supplier: G-Biosciences
Specifications
- Description:Size: 50 columns
- Environmentally Preferable:
- Pk:50
- Resin volume:3 ml
Specifications
About this item
The 3 ml spin columns are designed to hold up to 3 ml of resin, leaving enough rome for a further 4 ml of buffer. The unique design allows the columns to utilise a 15 ml centrifuge tube as a collection tube, so the columns can be centrifuged for rapid purification.
The columns can also be used for gravity flow purification.
Delivery: Supplied with matching cap, frits, and rubber stoppers installed.