Heat Resistant Gloves, A590
Supplier: Portwest
Certificates
About this item
The heat resistant gloves are constructed from Cotton/viscose material, which with a cotton liner, offers a long lasting gloves which is heat resistant to contact heat up to 250 °C. It is seamless, ambidextrous and machine washable.
- Provides contact heat protection up to 250 °C for 15 seconds
- Robust 7 gauge liner
- Long cuff version
- Ambidextrous and are suitable for left or right handed users for added functionality
- Sold in singles
- Machine washable to 30 °C
- Retail tag which aids presentation for retail sales
The Heat Protection range provides hand protection against temperatures ranging from 100 to 500 degrees for contact heat, while maintaining exceptional hand dexterity. Trusted to safeguard hands against heat hazards in all work environments.
Certifications: EN ISO 21420: 2020 Dexterity 1. EN 388: 2016 + A1: 2018 (1341X). EN 407: 2020 (4231XX).
Specifications
- Catalog No:
- PRTWA590WHRL
- PRTWA590WHRXXL
- Model:
- A590
- A590
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Liner material:
- Cotton
- Cotton
- Material:
- Cotton/viscose
- Cotton/viscose
- Cuff style:
- Long
- Long
Specifications
Frequently Bought Together