Order Entry
Ireland
ContactUsLinkComponent
Aluminium Twist Lock Carabiner, FP32
Aluminium Twist Lock Carabiner, FP32
Catalog # PRTWFP32SIR
Supplier:  Portwest
Aluminium Twist Lock Carabiner, FP32
Catalog # PRTWFP32SIR
Supplier:  Portwest

Specifications

  • Description:
    Height safety aluminium twist lock carabiner, FP32
  • Pk:
    1

Specifications

About this item

This lightweight twist-lock carabiner features a 20 mm gate for securing a connecting device such as a fall arrest lanyard to an anchorage point. Breaking load:23 kN.

  • Aluminium
  • Max rated load 100 kg
  • 10 Year shelf life from manufacturing date outlined in product label

Assort range of fall protection equipment to be used with our other fall protection equipment.

Height safety is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas. Portwest offer a full range of height safety products for working at height.

Certifications: CE certified. EN 362:2004 Class B, UKCA marked.