AlphaTec® 38-001 Butyl Gloves
Supplier: Ansell
Certificates
About this item
Delivering a high level of comfort and chemical protection, AlphaTec® 38-001 Series Butyl gloves offer superior protection against highly aggressive chemicals.
- Extended forearm protection
- Excellent dry and wet grip enhance dexterity and reduce hand fatigue
- Wide cuff to allow donning over protection clothing
AlphaTec® 38-001 Series Butyl gloves offer superior protection against highly aggressive chemicals, including aldehydes, ketones, esters, and concentrated mineral acids. Designed for comfort, these industrial gloves feature a unique butyl formulation to facilatate a soft feel, protecting hands while facilitating donning and doffing. Offering enhanced flexibility, the ultra-soft butyl formulation maximises flexibility and dexterity. The gloves also feature a raised diamond pattern, enhancing grip and ensuring safer handling.
Certifications: EN ISO 374-1:2016 Type A (ABCIKMPST), EN ISO 374-5 VIRUS, EN 421
Specifications
- Catalog No:
- ANSE38-001PP/10
- ANSE38-001PP/11
- ANSE38-001PP/9
- ANSE38-001PP/7
- ANSE38-001PP/8
- Model:
- 38-001
- 38-001
- 38-001
- 38-001
- 38-001
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Butyl
- Butyl
- Butyl
- Butyl
- Butyl
- Finish:
- Raised diamond grip
- Raised diamond grip
- Raised diamond grip
- Raised diamond grip
- Raised diamond grip
- Cuff style:
- Extended
- Extended
- Extended
- Extended
- Extended
- Palm thickness:
- 0,35 mm
- 0,35 mm
- 0,35 mm
- 0,35 mm
- 0,35 mm
- EN norm:
- EN ISO 374-1:2016 Type A (ABCIKMPST), EN ISO 374-5 VIRUS, EN 421
- EN ISO 374-1:2016 Type A (ABCIKMPST), EN ISO 374-5 VIRUS, EN 421
- EN ISO 374-1:2016 Type A (ABCIKMPST), EN ISO 374-5 VIRUS, EN 421
- EN ISO 374-1:2016 Type A (ABCIKMPST), EN ISO 374-5 VIRUS, EN 421
- EN ISO 374-1:2016 Type A (ABCIKMPST), EN ISO 374-5 VIRUS, EN 421
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile