Order Entry
Ireland
ContactUsLinkComponent
Recombinant Bovine IGF1
Recombinant Bovine IGF1
Catalog # ABNOP7663.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Bovine IGF1
Catalog # ABNOP7663.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Bovine
  • Gene ID:
    P07455
  • Protein/peptide name:
    IGF1
  • Sequence:
    50a.a.-119a.a.

Specifications

About this item