Recombinant Human RAET1L (from HEK293 cells), hFc tag
Catalog # ABNOP9710.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:hFc tag
- Gene ID:Q5VY80
- Protein/peptide name:RAET1L
- Sequence:26a.a.-218a.a.