Recombinant Human LGR5 (from HEK293 cells), hFc tag
Catalog # ABNOP9744.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:hFc tag
- Gene ID:O75473-1
- Protein/peptide name:LGR5
- Sequence:22a.a.-543a.a.