Order Entry
Ireland
ContactUsLinkComponent
Recombinant Human CCR8 (from HEK293 cells), hFc tag
Recombinant Human CCR8 (from HEK293 cells), hFc tag
  ABNOP9721.100UG
 :  Abnova
 :  
Recombinant Human CCR8 (from HEK293 cells), hFc tag
  ABNOP9721.100UG
 :  Abnova
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    hFc tag
  • Gene ID:
    P51685-1
  • Protein/peptide name:
    CCR8
  • Sequence:
    1a.a.-35a.a.