Recombinant Human NOTCH3 (from HEK293 cells), hFc tag
Catalog # ABNOP9726.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:hFc tag
- Gene ID:Q9UM47
- Protein/peptide name:NOTCH3
- Sequence:1378a.a.-1640a.a.