Order Entry
Ireland
ContactUsLinkComponent
 
Recombinant Human CD96 (from HEK293 cells), His-tag
Catalog # ABNOP9731.100UG
Supplier:  Abnova
CAS Number:  
Recombinant Human CD96 (from HEK293 cells), His-tag
Catalog # ABNOP9731.100UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    P40200-2
  • Protein/peptide name:
    CD96
  • Sequence:
    22a.a.-503a.a.

Specifications

About this item