Order Entry
Ireland
ContactUsLinkComponent
Recombinant Human F3 (from HEK293 cells), His-tag
Recombinant Human F3 (from HEK293 cells), His-tag
Catalog # ABNOP9737.100UG
Supplier:  Abnova
CAS Number:  
Recombinant Human F3 (from HEK293 cells), His-tag
Catalog # ABNOP9737.100UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    P13726-1
  • Protein/peptide name:
    F3
  • Sequence:
    33a.a.-251a.a.

Specifications

About this item