Recombinant Mouse CX3CL1 (from E. coli)
Catalog # ABNOP9456.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Gene ID:O35188
- Protein/peptide name:CX3CL1
- Sequence:25a.a.-100a.a.