Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human CSF3 (from CHO cells)
Recombinant Human CSF3 (from CHO cells)
Recombinant Human CSF3 (from CHO cells)
Catalog # ABNOP8705.10UG
Supplier:
Abnova
CAS Number:
Expand image
Recombinant Human CSF3 (from CHO cells)
Catalog # ABNOP8705.10UG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
10 µG
Protein/peptide type:
Recombinant
Source:
CHO cells
Species:
Human
Protein/peptide name:
CSF3
Specifications
About this item
Product Details
Product Details