Order Entry
Ireland
ContactUsLinkComponent
Recombinant Rat CSF3 (from E. coli)
Recombinant Rat CSF3 (from E. coli)
Catalog # ABNOP8709.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Rat CSF3 (from E. coli)
Catalog # ABNOP8709.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Protein/peptide name:
    CSF3

Specifications

About this item