Recombinant Horse IL2 (from E. coli)
Catalog # ABNOP8247.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Horse
- Gene ID:P37997
- Protein/peptide name:IL2
- Sequence:21a.a.-149a.a.