Recombinant Mouse IL6 (from E. coli)
Catalog # ABNOP8290.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Gene ID:P08505
- Protein/peptide name:IL6
- Sequence:26a.a.-211a.a.