Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human LGALS9 (from E. coli), His-tag
Recombinant Human LGALS9 (from E. coli), His-tag
ABNOP8676.10UG
:
Abnova
:
Recombinant Human LGALS9 (from E. coli), His-tag
ABNOP8676.10UG
:
Abnova
:
Pk:
10 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Tag sequence:
His-tag
Protein/peptide name:
LGALS9