Recombinant Human GFRA3 (from Baculovirus), His-tag
Catalog # ABNOP8688.5UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Human
- Tag sequence:His-tag
- Protein/peptide name:GFRA3