Order Entry
Ireland
ContactUsLinkComponent
 
Recombinant Rat IL17A (from E. coli)
Catalog # ABNOP8775.25UG
Supplier:  Abnova
CAS Number:  
Recombinant Rat IL17A (from E. coli)
Catalog # ABNOP8775.25UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Gene ID:
    Q61453
  • Protein/peptide name:
    IL17A

Specifications

About this item