Recombinant Human IL1B (from E. coli)
Catalog # ABNOP8123.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:P01584
- Protein/peptide name:IL1B
- Sequence:117a.a.-269a.a.