Recombinant Human BACE1 (from Baculovirus), His-tag
Catalog # ABNOP8066.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Human
- Tag sequence:His-tag
- Gene ID:P56817
- Protein/peptide name:BACE1
- Sequence:22a.a.-457a.a.