Recombinant Human PTPN11 (from Baculovirus), His-tag
ABNOP8073.100UG
: Abnova
:
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Human
- Tag sequence:His-tag
- Gene ID:Q06124
- Protein/peptide name:PTPN11
- Sequence:1a.a.-593a.a.