Order Entry
Ireland
ContactUsLinkComponent
Recombinant Mouse EPO (from Baculovirus), His-tag
Recombinant Mouse EPO (from Baculovirus), His-tag
Catalog # ABNOP8079.100UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse EPO (from Baculovirus), His-tag
Catalog # ABNOP8079.100UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Mouse
  • Tag sequence:
    His-tag
  • Gene ID:
    P07321
  • Protein/peptide name:
    EPO
  • Sequence:
    27a.a.-192a.a.

Specifications

About this item