Recombinant Human SLAMF7, His-Avi-tag
Catalog # ABNOP7657.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Tag sequence:His-Avi tag
- Gene ID:Q9NQ25
- Protein/peptide name:SLAMF7
- Sequence:23a.a.-226a.a.