Order Entry
Ireland
ContactUsLinkComponent
Recombinant Human IL15RA (from Baculovirus), hIgG-His-tag
Recombinant Human IL15RA (from Baculovirus), hIgG-His-tag
Catalog # ABNOP8028.250UG
Supplier:  Abnova
CAS Number:  
Recombinant Human IL15RA (from Baculovirus), hIgG-His-tag
Catalog # ABNOP8028.250UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    250 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Human
  • Tag sequence:
    hIgG-His-tag
  • Gene ID:
    Q13261
  • Protein/peptide name:
    IL15RA
  • Sequence:
    31a.a.-205a.a.

Specifications

About this item