Order Entry
Ireland
ContactUsLinkComponent
 
Recombinant Mouse UGDH (from E. coli), His-tag
Catalog # ABNOP8038.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse UGDH (from E. coli), His-tag
Catalog # ABNOP8038.20UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Tag sequence:
    His-tag
  • Gene ID:
    O70475
  • Protein/peptide name:
    UGDH
  • Sequence:
    1a.a.-493a.a.

Specifications

About this item