Recombinant Human IL21 (from Baculovirus), His-tag
Catalog # ABNOP8050.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Human
- Tag sequence:His-tag
- Gene ID:Q9HBE4
- Protein/peptide name:IL21
- Sequence:30a.a.-162a.a.