Recombinant Mouse CHST5 (from Baculovirus), His-tag
Catalog # ABNOP8095.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Mouse
- Tag sequence:His-tag
- Gene ID:Q9QUP4
- Protein/peptide name:CHST5
- Sequence:27a.a.-395a.a.