Recombinant Mouse TNFSF4 (from Baculovirus), His-tag
Catalog # ABNOP8106.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Mouse
- Tag sequence:His-tag
- Gene ID:P43488
- Protein/peptide name:TNFSF4
- Sequence:51a.a.-198a.a.