Recombinant Human SPON1 (from HEK293 cells), His-tag
ABNOP7910.50UG
: Abnova
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His-tag
- Gene ID:Q9HCB6
- Protein/peptide name:SPON1
- Sequence:29a.a.-807a.a.