Order Entry
Ireland
ContactUsLinkComponent
Recombinant Human P4HB (from E. coli), His-tag
Recombinant Human P4HB (from E. coli), His-tag
Catalog # ABNOP7937.100UG
Supplier:  Abnova
CAS Number:  
Recombinant Human P4HB (from E. coli), His-tag
Catalog # ABNOP7937.100UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    P07237
  • Protein/peptide name:
    P4HB
  • Sequence:
    18a.a.-508a.a.

Specifications

About this item