Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human NME2 (from E. coli)
Recombinant Human NME2 (from E. coli)
Recombinant Human NME2 (from E. coli)
ABNOP7962.100UG
:
Abnova
:
Recombinant Human NME2 (from E. coli)
ABNOP7962.100UG
:
Abnova
:
Pk:
100 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Gene ID:
P22392
Protein/peptide name:
NME2
Sequence:
1a.a.-152a.a.