0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Proteins and Peptides
Recombinant Human FGF19 (from E. coli), His-tag
Recombinant Human FGF19 (from E. coli), His-tag
Catalog # ABNOP7846.20UG
Supplier:
Abnova
CAS Number:
Expand image
Recombinant Human FGF19 (from E. coli), His-tag
Catalog # ABNOP7846.20UG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
20 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Tag sequence:
His-tag
Protein/peptide name:
FGF19
Specifications
About this item
Product Details
Product Details