Recombinant Human MSLN (from HEK293 cells), hFc tag
Catalog # ABNOP7650.1MG
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:hFc tag
- Gene ID:Q13421
- Protein/peptide name:MSLN
- Sequence:296a.a.-580a.a.