Order Entry
Ireland
ContactUsLinkComponent
Recombinant Human CDK2 (from Baculovirus), His-tag
Recombinant Human CDK2 (from Baculovirus), His-tag
  ABNOP7728.250UG
 :  Abnova
 :  
Recombinant Human CDK2 (from Baculovirus), His-tag
  ABNOP7728.250UG
 :  Abnova
 :  

 

  • Pk:
    250 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    NP_001789.2
  • Protein/peptide name:
    CDK2
  • Sequence:
    1a.a.-298a.a