Recombinant Human LCN1 (from E. coli), His-tag
Catalog # ABNOP7667.500UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:NP_001239546
- Protein/peptide name:LCN1
- Sequence:19a.a.-176a.a