Recombinant Human SNAPC1 (from E. coli), His-tag
ABNOP7668.500UG
: Abnova
:
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:NP_003073
- Protein/peptide name:SNAPC1
- Sequence:1a.a.-368a.a