Order Entry
Ireland
ContactUsLinkComponent
Recombinant Human MAK16 (from E. coli), His-tag
Recombinant Human MAK16 (from E. coli), His-tag
Catalog # ABNOP7671.250UG
Supplier:  Abnova
CAS Number:  
Recombinant Human MAK16 (from E. coli), His-tag
Catalog # ABNOP7671.250UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    250 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    NP_115898
  • Protein/peptide name:
    MAK16
  • Sequence:
    1a.a.-300a.a

Specifications

About this item