Order Entry
Ireland
ContactUsLinkComponent
 
Recombinant Human RAB39B (from E. coli), His-tag
Catalog # ABNOP7672.500UG
Supplier:  Abnova
CAS Number:  
Recombinant Human RAB39B (from E. coli), His-tag
Catalog # ABNOP7672.500UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    NP_741995.1
  • Protein/peptide name:
    RAB39B
  • Sequence:
    1a.a.-213a.a

Specifications

About this item