Recombinant Human ARFIP1 (from E. coli), His-tag
Catalog # ABNOP7674.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:NP_001020766
- Protein/peptide name:ARFIP1
- Sequence:1a.a.-373a.a