Order Entry
Ireland
ContactUsLinkComponent
 
Recombinant Human RPL35 (from E. coli), His-tag
  ABNOP7687.100UG
 :  Abnova
 :  
Recombinant Human RPL35 (from E. coli), His-tag
  ABNOP7687.100UG
 :  Abnova
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    NP_009140
  • Protein/peptide name:
    RPL35
  • Sequence:
    1a.a.-123a.a