Recombinant Human GAD1 (from E. coli), His-tag
ABNOP7689.500UG
: Abnova
:
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:NP_000808
- Protein/peptide name:GAD1
- Sequence:1a.a.-594a.a