Order Entry
Ireland
ContactUsLinkComponent
 
Recombinant Human CD37 (from E. coli), His-tag
  ABNOP7734.500UG
 :  Abnova
 :  
Recombinant Human CD37 (from E. coli), His-tag
  ABNOP7734.500UG
 :  Abnova
 :  

 

  • Pk:
    500 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    NP_001765
  • Protein/peptide name:
    CD37
  • Sequence:
    112a.a.-241a.a