Recombinant Human GTF3C6 (from E. coli), His-tag
Catalog # ABNOP7735.250UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:250 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:NP_612417
- Protein/peptide name:GTF3C6
- Sequence:1a.a.-213a.a