Recombinant Human PDCD1LG2 (from HEK293 cells), hFc tag
Catalog # ABNOP7568.1MG
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:hFc tag
- Gene ID:Q9BQ51
- Protein/peptide name:PDCD1LG2
- Sequence:20a.a.-219a.a.