Order Entry
Ireland
ContactUsLinkComponent
Recombinant Human TNFSF14 (from HEK293 cells)
Recombinant Human TNFSF14 (from HEK293 cells)
Catalog # ABNOP7612.50UG
Supplier:  Abnova
CAS Number:  
Recombinant Human TNFSF14 (from HEK293 cells)
Catalog # ABNOP7612.50UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Gene ID:
    O43557
  • Protein/peptide name:
    TNFSF14
  • Sequence:
    74a.a.-240a.a.

Specifications

About this item