Order Entry
Ireland
ContactUsLinkComponent
Recombinant Human FOLH1 (from CHO cells), hFc tag
Recombinant Human FOLH1 (from CHO cells), hFc tag
Catalog # ABNOP7614.50UG
Supplier:  Abnova
CAS Number:  
Recombinant Human FOLH1 (from CHO cells), hFc tag
Catalog # ABNOP7614.50UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Tag sequence:
    hFc tag
  • Gene ID:
    Q04609
  • Protein/peptide name:
    FOLH1
  • Sequence:
    44a.a.-750a.a.

Specifications

About this item