Order Entry
Ireland
ContactUsLinkComponent
Recombinant Human PDCD1 (from HEK293 cells), His-tag
Recombinant Human PDCD1 (from HEK293 cells), His-tag
Catalog # ABNOP7574.1MG
Supplier:  Abnova
CAS Number:  
Recombinant Human PDCD1 (from HEK293 cells), His-tag
Catalog # ABNOP7574.1MG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    Q15116
  • Protein/peptide name:
    PDCD1
  • Sequence:
    25a.a.-167a.a.

Specifications

About this item